Product Details

SNP ID
rs79329361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:9092965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACTTTCTCTCTGGGTCCCCTTC[A/C]AAAACGCAGCCACCGCCGTCACCAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C16orf72 PubMed Links

Gene Details

Gene
C16orf72
Gene Name
chromosome 16 open reading frame 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014117.2 636 Missense Mutation AAA,CAA K91Q NP_054836.2
XM_011522462.2 636 Missense Mutation AAA,CAA K91Q XP_011520764.1

View Full Product Details