Product Details

SNP ID
rs79818496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:9754171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAATATATATACTATACATAAA[C/G]ATATACACATAAACATTCTGGGGTG
Phenotype
MIM: 138253
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GRIN2A PubMed Links

Gene Details

Gene
GRIN2A
Gene Name
glutamate ionotropic receptor NMDA type subunit 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000833.4 13169 UTR 3 NP_000824.1
NM_001134407.2 13169 UTR 3 NP_001127879.1
NM_001134408.2 13169 UTR 3 NP_001127880.1
XM_011522458.2 13169 UTR 3 XP_011520760.1
XM_011522461.2 13169 UTR 3 XP_011520763.1
XM_017023172.1 13169 UTR 3 XP_016878661.1
XM_017023173.1 13169 UTR 3 XP_016878662.1

View Full Product Details