Product Details

SNP ID
rs78289309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57698349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACTAGCTGTGTGACCTTGGGCA[A/C]GTCACTCAGCCTCTCGGTGCCTCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADGRG3 PubMed Links

Gene Details

Gene
ADGRG3
Gene Name
adhesion G protein-coupled receptor G3
There are no transcripts associated with this gene.

Gene
DRC7
Gene Name
dynein regulatory complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289162.1 Intron NP_001276091.1
NM_001289163.1 Intron NP_001276092.1
NM_032269.5 Intron NP_115645.4
XM_011523377.1 Intron XP_011521679.1
XM_011523378.1 Intron XP_011521680.1

View Full Product Details