Product Details

SNP ID
rs75245751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:61640974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGTGCTGATTTTTCTCTTCATT[A/C]ATGATTGTTAGCCTCACCAACTGTT
Phenotype
MIM: 603008
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDH8 PubMed Links

Gene Details

Gene
CDH8
Gene Name
cadherin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001796.4 9580 Intron NP_001787.2
XM_005255760.3 9580 UTR 3 XP_005255817.1
XM_017022829.1 9580 Intron XP_016878318.1

View Full Product Details