Product Details

SNP ID
rs77744771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:48247946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCTGGCCTATGTTTATAATTCT[C/T]GTAGGTAGAAGTGGTACCTATTGTC
Phenotype
MIM: 607040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCC11 PubMed Links

Gene Details

Gene
ABCC11
Gene Name
ATP binding cassette subfamily C member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032583.3 2028 Intron NP_115972.2
NM_033151.3 2028 Intron NP_149163.2
NM_145186.2 2028 Intron NP_660187.1
XM_011523397.1 2028 Intron XP_011521699.1
XM_011523398.2 2028 Intron XP_011521700.1
XM_017023795.1 2028 UTR 5 XP_016879284.1
XM_017023796.1 2028 Intron XP_016879285.1
XM_017023797.1 2028 Intron XP_016879286.1
XM_017023798.1 2028 Intron XP_016879287.1
XM_017023799.1 2028 Intron XP_016879288.1
XM_017023800.1 2028 Intron XP_016879289.1
XM_017023801.1 2028 UTR 5 XP_016879290.1
XM_017023802.1 2028 Intron XP_016879291.1
XM_017023803.1 2028 UTR 5 XP_016879292.1
Gene
LONP2
Gene Name
lon peptidase 2, peroxisomal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300948.1 2028 Intron NP_001287877.1
NM_031490.3 2028 Intron NP_113678.2
XM_017023755.1 2028 Intron XP_016879244.1
XM_017023756.1 2028 Intron XP_016879245.1

View Full Product Details