Product Details

SNP ID
rs79985162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66884464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCATGTGGTGGCTTTACTCTGTG[A/C]AAAGCCTACAGACACACATCAACAG
Phenotype
MIM: 615499
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PDP2 PubMed Links

Gene Details

Gene
PDP2
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005256064.4 509 Nonsense Mutation TGA,TGC *60C XP_005256121.1
XM_005256066.4 509 Nonsense Mutation TGA,TGC *60C XP_005256123.1
XM_005256067.4 509 Nonsense Mutation TGA,TGC *60C XP_005256124.1
XM_005256068.4 509 Nonsense Mutation TGA,TGC *60C XP_005256125.1
XM_017023506.1 509 Nonsense Mutation TGA,TGC *60C XP_016878995.1

View Full Product Details