Product Details

SNP ID
rs77700569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70807305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAAATGGGAATTATTACAAAGT[A/C]CTTGAGCTTTGGGGCTATGTCAAGA
Phenotype
MIM: 610812 MIM: 604632
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HYDIN PubMed Links

Gene Details

Gene
HYDIN
Gene Name
HYDIN, axonemal central pair apparatus protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198542.1 15854 Intron NP_001185471.1
NM_001198543.1 15854 Intron NP_001185472.1
NM_001270974.2 15854 UTR 3 NP_001257903.1
NM_017558.4 15854 Intron NP_060028.2
XM_006721206.2 15854 UTR 3 XP_006721269.1
XM_011523146.2 15854 UTR 3 XP_011521448.1
XM_011523147.1 15854 UTR 3 XP_011521449.1
XM_011523148.1 15854 UTR 3 XP_011521450.1
XM_011523151.1 15854 UTR 3 XP_011521453.1
XM_011523152.1 15854 UTR 3 XP_011521454.1
XM_011523155.2 15854 UTR 3 XP_011521457.1
XM_017023346.1 15854 UTR 3 XP_016878835.1
XM_017023347.1 15854 UTR 3 XP_016878836.1
XM_017023348.1 15854 UTR 3 XP_016878837.1
Gene
VAC14
Gene Name
Vac14, PIKFYVE complex component
There are no transcripts associated with this gene.

View Full Product Details