Product Details

SNP ID
rs79926541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70810293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATAGGGAGATGTGGAGATGGGT[A/G]AGTGCTGCCAGTAGGAGGGCTGAAG
Phenotype
MIM: 610812 MIM: 604632
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HYDIN PubMed Links

Gene Details

Gene
HYDIN
Gene Name
HYDIN, axonemal central pair apparatus protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198542.1 Intron NP_001185471.1
NM_001198543.1 Intron NP_001185472.1
NM_001270974.2 Intron NP_001257903.1
NM_017558.4 Intron NP_060028.2
XM_006721206.2 Intron XP_006721269.1
XM_011523146.2 Intron XP_011521448.1
XM_011523147.1 Intron XP_011521449.1
XM_011523148.1 Intron XP_011521450.1
XM_011523151.1 Intron XP_011521453.1
XM_011523152.1 Intron XP_011521454.1
XM_011523155.2 Intron XP_011521457.1
XM_017023346.1 Intron XP_016878835.1
XM_017023347.1 Intron XP_016878836.1
XM_017023348.1 Intron XP_016878837.1
Gene
VAC14
Gene Name
Vac14, PIKFYVE complex component
There are no transcripts associated with this gene.

View Full Product Details