Product Details

SNP ID
rs78008482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75112615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCTCCTACCTGCCCAGCTGTT[C/T]TGCAAAAGCCTTGACCCTGCCCAGT
Phenotype
MIM: 607490 MIM: 612060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDHD PubMed Links

Gene Details

Gene
LDHD
Gene Name
lactate dehydrogenase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153486.3 1328 Missense Mutation AAA,GAA K449E NP_705690.2
NM_194436.2 1328 Missense Mutation AAA,GAA K426E NP_919417.1
Gene
ZNRF1
Gene Name
zinc and ring finger 1, E3 ubiquitin protein ligase
There are no transcripts associated with this gene.

View Full Product Details