Product Details
- SNP ID
-
rs77436937
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75529857 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGGGCCAGGCTGAATGGCTGCCG[C/T]GTGCACAGTGTCTTGCATACGTCCT
- Phenotype
-
MIM: 604817
MIM: 614949
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHST5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8048818] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHST5
- Gene Name
- carbohydrate sulfotransferase 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024533.4 |
1923 |
Silent Mutation |
ACA,ACG |
T176T |
NP_078809.2 |
- Gene
- TMEM231
- Gene Name
- transmembrane protein 231
There are no transcripts associated with this gene.
View Full Product Details