Product Details

SNP ID
rs75930424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:80602169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCATCTAAGGGTTTTGGAAAGCT[C/T]ACATTTGCTTTCTCAAAGGAGGAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDYL2 PubMed Links

Gene Details

Gene
CDYL2
Gene Name
chromodomain protein, Y-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152342.3 4400 UTR 3 NP_689555.2
XM_011522866.1 4400 UTR 3 XP_011521168.1
XM_011522867.2 4400 UTR 3 XP_011521169.1

View Full Product Details