Product Details

SNP ID
rs74403414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81241444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAAAACATGAATTCTAGAACAC[A/G]AGAGGCTGAGTGAGCCCCAAAGATC
Phenotype
MIM: 605748
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCO1 PubMed Links

Gene Details

Gene
BCO1
Gene Name
beta-carotene oxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017429.2 Intron NP_059125.2
XM_011523109.1 Intron XP_011521411.1
XM_017023286.1 Intron XP_016878775.1
XM_017023287.1 Intron XP_016878776.1
XM_017023288.1 Intron XP_016878777.1
XM_017023289.1 Intron XP_016878778.1

View Full Product Details