Product Details

SNP ID
rs78018668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:86567907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAGGAGCCGCCCCCGGCGGCGT[C/T]CAAGGGCGCCCCGGCCACCCCCCAC
Phenotype
MIM: 602402
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXC2 PubMed Links

Gene Details

Gene
FOXC2
Gene Name
forkhead box C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005251.2 657 Missense Mutation TCC,TTC S191F NP_005242.1
Gene
FOXC2-AS1
Gene Name
FOXC2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details