Product Details

SNP ID
rs79124041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50668046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCGTCAGGAGCCGGAGCCTAC[A/G]GGTTGAAAGCCAAGAGTGGGCAATC
Phenotype
MIM: 613281
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927272 PubMed Links

Gene Details

Gene
LOC101927272
Gene Name
uncharacterized LOC101927272
There are no transcripts associated with this gene.

Gene
SNX20
Gene Name
sorting nexin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144972.1 1558 Intron NP_001138444.1
NM_153337.2 1558 UTR 3 NP_699168.1
NM_182854.3 1558 Intron NP_878274.1

View Full Product Details