Product Details

SNP ID
rs77286977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:47083686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCATGACCTTTTTTCGAGGCTG[G/T]TTCACTTGTACTAAAATAGAAATAA
Phenotype
MIM: 607974
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NETO2 PubMed Links

Gene Details

Gene
NETO2
Gene Name
neuropilin and tolloid like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201477.1 1505 Missense Mutation AAA,AAC K364N NP_001188406.1
NM_018092.4 1505 Missense Mutation AAA,AAC K371N NP_060562.3
XM_006721289.1 1505 Missense Mutation AAA,AAC K333N XP_006721352.1
XM_006721291.1 1505 Missense Mutation AAA,AAC K326N XP_006721354.1
XM_006721292.1 1505 Intron XP_006721355.1
XM_017023740.1 1505 Missense Mutation AAA,AAC K235N XP_016879229.1

View Full Product Details