Product Details

SNP ID
rs76888998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89817902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTGCAATCTGCTGGTTGAGGG[G/T]GATGGTGTGAACTCAGGGAACAGCC
Phenotype
MIM: 607139 MIM: 609217
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FANCA PubMed Links

Gene Details

Gene
FANCA
Gene Name
Fanconi anemia complementation group A
There are no transcripts associated with this gene.

Gene
SPIRE2
Gene Name
spire type actin nucleation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032451.1 426 Intron NP_115827.1
XM_011523379.1 426 UTR 5 XP_011521681.1
XM_011523380.2 426 Intron XP_011521682.1
XM_011523381.1 426 UTR 5 XP_011521683.1
XM_011523384.1 426 Intron XP_011521686.1
XM_011523385.1 426 Intron XP_011521687.1
XM_017023779.1 426 Intron XP_016879268.1
XM_017023780.1 426 Intron XP_016879269.1
XM_017023781.1 426 Intron XP_016879270.1

View Full Product Details