Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014972.2 | Intron | NP_055787.1 | |||
XM_005256297.2 | Intron | XP_005256354.1 | |||
XM_005256298.2 | Intron | XP_005256355.1 | |||
XM_005256299.2 | Intron | XP_005256356.1 | |||
XM_011522956.2 | Intron | XP_011521258.1 | |||
XM_011522957.2 | Intron | XP_011521259.1 | |||
XM_011522958.2 | Intron | XP_011521260.1 | |||
XM_011522959.2 | Intron | XP_011521261.1 | |||
XM_011522960.2 | Intron | XP_011521262.1 | |||
XM_011522961.2 | Intron | XP_011521263.1 | |||
XM_017023047.1 | Intron | XP_016878536.1 | |||
XM_017023048.1 | Intron | XP_016878537.1 | |||
XM_017023049.1 | Intron | XP_016878538.1 | |||
XM_017023050.1 | Intron | XP_016878539.1 | |||
XM_017023051.1 | Intron | XP_016878540.1 | |||
XM_017023052.1 | Intron | XP_016878541.1 | |||
XM_017023053.1 | Intron | XP_016878542.1 | |||
XM_017023054.1 | Intron | XP_016878543.1 | |||
XM_017023055.1 | Intron | XP_016878544.1 | |||
XM_017023056.1 | Intron | XP_016878545.1 | |||
XM_017023057.1 | Intron | XP_016878546.1 | |||
XM_017023058.1 | Intron | XP_016878547.1 |