Product Details

SNP ID
rs77913543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10824644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGTCATGTTGTCATGTCCAGGG[G/T]CACGTGCCATCTTGGAGAGTATTTT
Phenotype
MIM: 612068
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PIRT PubMed Links

Gene Details

Gene
PIRT
Gene Name
phosphoinositide interacting regulator of transient receptor potential channels
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101387.1 1297 UTR 3 NP_001094857.1
Gene
TMEM220-AS1
Gene Name
TMEM220 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details