Product Details

SNP ID
rs75116322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:14075444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTCCCATGCCTGCCTATAGCTG[A/G]TTGTTGGCTGTTGATATATCAGTTT
Phenotype
MIM: 602125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX10 PubMed Links

Gene Details

Gene
COX10
Gene Name
COX10, heme A:farnesyltransferase cytochrome c oxidase assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303.3 Intron NP_001294.2
Gene
COX10-AS1
Gene Name
COX10 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details