Product Details

SNP ID
rs77724683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2315861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATACAGCTGGAAAAAAGGTTACA[A/G]AAATTTTCCTTTCAAGTCCTTGTTT
Phenotype
MIM: 606477 MIM: 611214
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRR PubMed Links

Gene Details

Gene
SRR
Gene Name
serine racemase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304803.1 Intron NP_001291732.1
NM_021947.2 Intron NP_068766.1
XM_006721565.3 Intron XP_006721628.1
XM_006721566.3 Intron XP_006721629.1
XM_011523974.2 Intron XP_011522276.1
Gene
TSR1
Gene Name
TSR1, ribosome maturation factor
There are no transcripts associated with this gene.

View Full Product Details