Product Details

SNP ID
rs74989743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4940489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTTTAACCGGCCTTTTAGGA[C/T]CGGAAGTCCTTCATCTCAAGCATCC
Phenotype
MIM: 131370 MIM: 606672 MIM: 176610 MIM: 610431 MIM: 604165
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENO3 PubMed Links

Gene Details

Gene
ENO3
Gene Name
enolase 3
There are no transcripts associated with this gene.

Gene
GP1BA
Gene Name
glycoprotein Ib platelet alpha subunit
There are no transcripts associated with this gene.

Gene
PFN1
Gene Name
profilin 1
There are no transcripts associated with this gene.

Gene
RNF167
Gene Name
ring finger protein 167
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320356.1 226 UTR 5 NP_001307285.1
NM_001320357.1 226 Intron NP_001307286.1
NM_001320358.1 226 Intron NP_001307287.1
NM_001320359.1 226 UTR 5 NP_001307288.1
NM_001320360.1 226 UTR 5 NP_001307289.1
NM_001320361.1 226 UTR 5 NP_001307290.1
NM_001320362.1 226 Intron NP_001307291.1
NM_001320363.1 226 Intron NP_001307292.1
NM_001320364.1 226 Intron NP_001307293.1
NM_001320365.1 226 UTR 5 NP_001307294.1
NM_015528.2 226 UTR 5 NP_056343.1
XM_017024427.1 226 UTR 5 XP_016879916.1
XM_017024428.1 226 Intron XP_016879917.1
Gene
SLC25A11
Gene Name
solute carrier family 25 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165417.1 226 Intron NP_001158889.1
NM_001165418.1 226 Intron NP_001158890.1
NM_003562.4 226 Intron NP_003553.2

View Full Product Details