Product Details

SNP ID
rs79167477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7421191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTCCCTGGTCTCAGGATGCCCC[A/G]GAGTCCCCTCCCCAGACCATCCGCT
Phenotype
MIM: 606479 MIM: 615116
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf74 PubMed Links

Gene Details

Gene
C17orf74
Gene Name
chromosome 17 open reading frame 74
There are no transcripts associated with this gene.

Gene
LOC107983988
Gene Name
proteoglycan 4-like
There are no transcripts associated with this gene.

Gene
NLGN2
Gene Name
neuroligin 2
There are no transcripts associated with this gene.

Gene
SPEM1
Gene Name
spermatid maturation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199339.2 541 Silent Mutation CCA,CCG P172P NP_955371.2

View Full Product Details