Product Details

SNP ID
rs76766056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47256618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGGGACTCCAAGTGTTGAAGGG[A/G]CACAGAAAGAGCTGAGTGGCACAGA
Phenotype
MIM: 173470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGB3 PubMed Links

Gene Details

Gene
ITGB3
Gene Name
integrin subunit beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000212.2 Intron NP_000203.2

View Full Product Details