Product Details

SNP ID
rs74333450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48766458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGCACTTTATCACACCCTCTT[A/C]ATATAATGATCTCTTCCCTTTCTTC
Phenotype
MIM: 610849
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TTLL6 PubMed Links

Gene Details

Gene
TTLL6
Gene Name
tubulin tyrosine ligase like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130918.1 Intron NP_001124390.1
NM_173623.3 Intron NP_775894.2
XM_017024491.1 Intron XP_016879980.1
XM_017024492.1 Intron XP_016879981.1
XM_017024493.1 Intron XP_016879982.1
XM_017024494.1 Intron XP_016879983.1
XM_017024495.1 Intron XP_016879984.1
XM_017024496.1 Intron XP_016879985.1
XM_017024497.1 Intron XP_016879986.1

View Full Product Details