Product Details

SNP ID
rs74923623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:51633574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCAGCGGTTGTTGAGTGGCTGG[A/G]CAGGCCTGAAGTTGTCACTCATGCT
Phenotype
MIM: 604642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CA10 PubMed Links

Gene Details

Gene
CA10
Gene Name
carbonic anhydrase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082533.1 1852 Missense Mutation GCC,GTC A289V NP_001076002.1
NM_001082534.1 1852 Missense Mutation GCC,GTC A289V NP_001076003.1
NM_020178.4 1852 Missense Mutation GCC,GTC A289V NP_064563.1
XM_017024878.1 1852 Missense Mutation GCC,GTC A192V XP_016880367.1

View Full Product Details