Product Details

SNP ID
rs75358646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:54906880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTGATTTATTCTGCACGCCTT[C/T]GTTTATCTCAGCCCAGCCCAAGACT
Phenotype
MIM: 604701
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOM1L1 PubMed Links

Gene Details

Gene
TOM1L1
Gene Name
target of myb1 like 1 membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321173.1 Intron NP_001308102.1
NM_001321174.1 Intron NP_001308103.1
NM_001321175.1 Intron NP_001308104.1
NM_001321176.1 Intron NP_001308105.1
NM_005486.2 Intron NP_005477.2
XM_017024002.1 Intron XP_016879491.1

View Full Product Details