Product Details

SNP ID
rs78936308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58169858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTTCTGCTCCGAAACCTGGCTG[G/T]CCTAGACCTCTGTTTCTCTTCAGTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4D2 PubMed Links

Gene Details

Gene
OR4D2
Gene Name
olfactory receptor family 4 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004707.3 203 Missense Mutation GGC,GTC G68V NP_001004707.1

View Full Product Details