Product Details

SNP ID
rs74324354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38735499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGCTGACTGACTCACACTCGGA[C/T]GCCCCGCTGGTGTTGGTGCCCTCGG
Phenotype
MIM: 611933 MIM: 600328 MIM: 600346
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CISD3 PubMed Links

Gene Details

Gene
CISD3
Gene Name
CDGSH iron sulfur domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136498.1 2021 UTR 3 NP_001129970.1
Gene
MLLT6
Gene Name
MLLT6, PHD finger domain containing
There are no transcripts associated with this gene.

Gene
PCGF2
Gene Name
polycomb group ring finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007144.2 2021 Silent Mutation GCA,GCG A253A NP_009075.1
XM_005257640.2 2021 Silent Mutation GCA,GCG A253A XP_005257697.1
XM_005257641.4 2021 Silent Mutation GCA,GCG A253A XP_005257698.1
XM_005257642.3 2021 Silent Mutation GCA,GCG A253A XP_005257699.1
XM_017025016.1 2021 Silent Mutation GCA,GCG A253A XP_016880505.1

View Full Product Details