Product Details

SNP ID
rs75172605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38767596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTCGTCTTTAACCAGTAAGAAT[A/G]TGTTACACTTTCCATACACATTATG
Phenotype
MIM: 603261 MIM: 602176
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIP4K2B PubMed Links

Gene Details

Gene
PIP4K2B
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003559.4 3790 UTR 3 NP_003550.1
XM_011525326.2 3790 UTR 3 XP_011523628.1
XM_011525327.2 3790 UTR 3 XP_011523629.1
XM_011525329.2 3790 Intron XP_011523631.1
XM_011525330.2 3790 UTR 3 XP_011523632.1
XM_017025196.1 3790 UTR 3 XP_016880685.1
XM_017025197.1 3790 UTR 3 XP_016880686.1
XM_017025198.1 3790 UTR 3 XP_016880687.1
XM_017025199.1 3790 UTR 3 XP_016880688.1
Gene
PSMB3
Gene Name
proteasome subunit beta 3
There are no transcripts associated with this gene.

View Full Product Details