Product Details

SNP ID
rs80316873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59896566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTCTTTTCACTTTCTTGTGTT[G/T]CAAGAACAGTAGGTGTTCCAGGGCT
Phenotype
MIM: 608938 MIM: 607344
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RPS6KB1 PubMed Links

Gene Details

Gene
RPS6KB1
Gene Name
ribosomal protein S6 kinase B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272042.1 Intron NP_001258971.1
NM_001272043.1 Intron NP_001258972.1
NM_001272044.1 Intron NP_001258973.1
NM_001272060.1 Intron NP_001258989.1
NM_003161.3 Intron NP_003152.1
XM_011525101.2 Intron XP_011523403.1
XM_011525102.2 Intron XP_011523404.1
XM_011525103.2 Intron XP_011523405.1
XM_017024929.1 Intron XP_016880418.1
XM_017024930.1 Intron XP_016880419.1
XM_017024931.1 Intron XP_016880420.1
XM_017024932.1 Intron XP_016880421.1
XM_017024933.1 Intron XP_016880422.1
Gene
TUBD1
Gene Name
tubulin delta 1
There are no transcripts associated with this gene.

View Full Product Details