Product Details

SNP ID
rs79294667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63557092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTAGAAAGATGCTATTAGAAAG[A/G]ATGGGGGGAAATGGAAAATAAGTGG
Phenotype
MIM: 605973 MIM: 608168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF7 PubMed Links

Gene Details

Gene
DCAF7
Gene Name
DDB1 and CUL4 associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005828.4 Intron NP_005819.3
Gene
KCNH6
Gene Name
potassium voltage-gated channel subfamily H member 6
There are no transcripts associated with this gene.

View Full Product Details