Product Details

SNP ID
rs75264656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63626777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGGGACACTGAAGCACAGACT[C/T]TCCTGAGTGGGCAGTGCGATCAGAC
Phenotype
MIM: 602539
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP3K3 PubMed Links

Gene Details

Gene
MAP3K3
Gene Name
mitogen-activated protein kinase kinase kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002401.3 Intron NP_002392.2
NM_203351.1 Intron NP_976226.1
XM_005257376.3 Intron XP_005257433.1
XM_005257377.3 Intron XP_005257434.1
XM_005257378.2 Intron XP_005257435.1

View Full Product Details