Product Details

SNP ID
rs74401893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67720753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAAACTAGGTTAATCTTGAAAGT[C/T]CTTTCCAGTTCTAAATGTCTAAATA
Phenotype
MIM: 615366
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOL11 PubMed Links

Gene Details

Gene
NOL11
Gene Name
nucleolar protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303272.1 Intron NP_001290201.1
NM_015462.4 Intron NP_056277.2

View Full Product Details