Product Details

SNP ID
rs77077922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40132065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGAGGGACCACTCAGTAGAGCC[C/T]CTAAGGGACCCAAATCCTTCAGACC
Phenotype
MIM: 606504 MIM: 614801
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASC3 PubMed Links

Gene Details

Gene
CASC3
Gene Name
cancer susceptibility candidate 3
There are no transcripts associated with this gene.

Gene
MSL1
Gene Name
male specific lethal 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012241.1 1007 Silent Mutation CCC,CCT P222P NP_001012241.1
XM_005257298.4 1007 Silent Mutation CCC,CCT P485P XP_005257355.1
XM_005257299.4 1007 Silent Mutation CCC,CCT P469P XP_005257356.3

View Full Product Details