Product Details

SNP ID
rs75957101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40141732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTGTGTATTCTTCATTTATGC[G/T]TCTGAAAAAGTGTTACAGAAGGTCT
Phenotype
MIM: 606504 MIM: 614801
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CASC3 PubMed Links

Gene Details

Gene
CASC3
Gene Name
cancer susceptibility candidate 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007359.4 Intron NP_031385.2
XM_005257163.1 Intron XP_005257220.1
Gene
MSL1
Gene Name
male specific lethal 1 homolog
There are no transcripts associated with this gene.

View Full Product Details