Product Details
- SNP ID
-
rs75283233
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:80467895 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTATCAAAAGCAAGAAGGCTTA[C/T]GACTCAAGTGGGATCTAAGCCCCTG
- Phenotype
-
MIM: 602367
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NPTX1
PubMed Links
Gene Details
- Gene
- NPTX1
- Gene Name
- neuronal pentraxin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002522.3 |
4375 |
UTR 3 |
|
|
NP_002513.2 |
View Full Product Details