Product Details

SNP ID
rs79037131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37087049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACATGTTTGTACCTTTCATTGC[C/T]TTCTCAGTCCTGTGCAGGGAGTGGA
Phenotype
MIM: 200350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACACA PubMed Links

Gene Details

Gene
ACACA
Gene Name
acetyl-CoA carboxylase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198834.2 8605 UTR 3 NP_942131.1
NM_198836.2 8605 UTR 3 NP_942133.1
NM_198837.1 8605 UTR 3 NP_942134.1
NM_198838.1 8605 UTR 3 NP_942135.1
NM_198839.2 8605 UTR 3 NP_942136.1
XM_005257267.4 8605 UTR 3 XP_005257324.1
XM_006721853.1 8605 UTR 3 XP_006721916.1
XM_011524701.1 8605 UTR 3 XP_011523003.1
XM_011524703.1 8605 UTR 3 XP_011523005.1
XM_011524704.2 8605 UTR 3 XP_011523006.1
XM_017024553.1 8605 UTR 3 XP_016880042.1
XM_017024554.1 8605 UTR 3 XP_016880043.1
XM_017024555.1 8605 UTR 3 XP_016880044.1

View Full Product Details