Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001098833.1 | 2948 | Intron | NP_001092303.1 | ||
NM_020218.1 | 2948 | Intron | NP_064603.1 | ||
XM_011525038.2 | 2948 | Intron | XP_011523340.1 | ||
XM_011525044.2 | 2948 | Intron | XP_011523346.1 | ||
XM_011525046.2 | 2948 | Intron | XP_011523348.1 | ||
XM_017024880.1 | 2948 | UTR 3 | XP_016880369.1 | ||
XM_017024881.1 | 2948 | UTR 3 | XP_016880370.1 | ||
XM_017024882.1 | 2948 | Intron | XP_016880371.1 | ||
XM_017024883.1 | 2948 | UTR 3 | XP_016880372.1 | ||
XM_017024884.1 | 2948 | UTR 3 | XP_016880373.1 | ||
XM_017024885.1 | 2948 | UTR 3 | XP_016880374.1 | ||
XM_017024886.1 | 2948 | UTR 3 | XP_016880375.1 |