Product Details

SNP ID
rs75193644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28347485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCCCTCCTCTCCTGGGCCTAGC[A/G]AGGCCAGAAATGGCAATGGTCGGTC
Phenotype
MIM: 611519 MIM: 191161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLDIP2 PubMed Links

Gene Details

Gene
POLDIP2
Gene Name
DNA polymerase delta interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290145.1 1826 UTR 3 NP_001277074.1
NM_015584.4 1826 UTR 3 NP_056399.1
Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021137.4 1826 Intron NP_066960.1
XM_017024993.1 1826 Intron XP_016880482.1

View Full Product Details