Product Details

SNP ID
rs77056680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34961636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAAATCAGGAAGAATTGCGGCGG[G/T]TAATGAAGGAGAAGCAGCGTCTGAG
Phenotype
MIM: 610730
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCT6B PubMed Links

Gene Details

Gene
CCT6B
Gene Name
chaperonin containing TCP1 subunit 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193529.2 107 Intron NP_001180458.1
NM_001193530.1 107 Intron NP_001180459.1
NM_006584.3 107 Intron NP_006575.2
XM_011524203.2 107 Intron XP_011522505.1
XM_017024023.1 107 Intron XP_016879512.1
XM_017024024.1 107 Intron XP_016879513.1
Gene
ZNF830
Gene Name
zinc finger protein 830
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052857.3 107 Missense Mutation GTA,TTA V24L NP_443089.3

View Full Product Details