Product Details

SNP ID
rs78518141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35760491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAGGATAAGGATAAGACAGGA[A/G]GAGGAAGACAGAATATAATGATACA
Phenotype
MIM: 611398 MIM: 608417
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf50 PubMed Links

Gene Details

Gene
C17orf50
Gene Name
chromosome 17 open reading frame 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145272.3 Intron NP_660315.2
Gene
GAS2L2
Gene Name
growth arrest specific 2 like 2
There are no transcripts associated with this gene.

Gene
MMP28
Gene Name
matrix metallopeptidase 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032278.2 Intron NP_001027449.1
NM_024302.4 Intron NP_077278.1
NM_032950.3 Intron NP_116568.1
XM_011525225.1 Intron XP_011523527.1
XM_011525226.2 Intron XP_011523528.1
XM_011525227.1 Intron XP_011523529.1
XM_011525228.1 Intron XP_011523530.1
XM_011525229.2 Intron XP_011523531.1
XM_011525230.1 Intron XP_011523532.1
XM_011525231.1 Intron XP_011523533.1
XM_011525232.2 Intron XP_011523534.2
XM_017025061.1 Intron XP_016880550.1
XM_017025062.1 Intron XP_016880551.1
XM_017025063.1 Intron XP_016880552.1
XM_017025064.1 Intron XP_016880553.1

View Full Product Details