Product Details

SNP ID
rs78564115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:8707355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGGGCCAGTCCTTTGTAAGCAT[C/T]CAGGAGCCGGCTTTCTGTGTCAGGA
Phenotype
MIM: 616131 MIM: 615766
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GACAT2 PubMed Links

Gene Details

Gene
GACAT2
Gene Name
gastric cancer associated transcript 2 (non-protein coding)
There are no transcripts associated with this gene.

Gene
MTCL1
Gene Name
microtubule crosslinking factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015210.3 Intron NP_056025.2
XM_005258098.3 Intron XP_005258155.1
XM_005258099.4 Intron XP_005258156.1
XM_011525640.2 Intron XP_011523942.1
XM_011525641.2 Intron XP_011523943.1
XM_017025671.1 Intron XP_016881160.1
XM_017025672.1 Intron XP_016881161.1
XM_017025673.1 Intron XP_016881162.1
XM_017025674.1 Intron XP_016881163.1
XM_017025675.1 Intron XP_016881164.1

View Full Product Details