Product Details

SNP ID
rs78004015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:45985387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTGCTGACATAACGTGGCTATA[C/G]GTCCTGCTGCTCTGGGTGCCCTTTC
Phenotype
MIM: 616046
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PSTPIP2 PubMed Links

Gene Details

Gene
PSTPIP2
Gene Name
proline-serine-threonine phosphatase interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024430.3 1050 UTR 3 NP_077748.3
XM_011526253.2 1050 UTR 3 XP_011524555.1
XM_011526254.1 1050 UTR 3 XP_011524556.1
XM_011526256.1 1050 UTR 3 XP_011524558.1

View Full Product Details