Product Details

SNP ID
rs78773539
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:48027720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGCTGAAGTCTCTTTGTGCCCA[C/T]AGCACAGGGATTGGGGACTCCAGCC
Phenotype
MIM: 616591
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZBTB7C PubMed Links

Gene Details

Gene
ZBTB7C
Gene Name
zinc finger and BTB domain containing 7C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039360.2 4113 UTR 3 NP_001034449.1
NM_001318841.1 4113 UTR 3 NP_001305770.1
XM_005258229.4 4113 UTR 3 XP_005258286.1
XM_011525861.2 4113 UTR 3 XP_011524163.1
XM_011525863.2 4113 UTR 3 XP_011524165.1
XM_011525864.2 4113 UTR 3 XP_011524166.1
XM_011525865.2 4113 UTR 3 XP_011524167.1
XM_011525866.2 4113 UTR 3 XP_011524168.1
XM_011525867.2 4113 UTR 3 XP_011524169.1
XM_011525869.2 4113 UTR 3 XP_011524171.1
XM_011525870.2 4113 UTR 3 XP_011524172.1
XM_011525871.2 4113 UTR 3 XP_011524173.1
XM_017025605.1 4113 UTR 3 XP_016881094.1
XM_017025606.1 4113 UTR 3 XP_016881095.1
XM_017025607.1 4113 UTR 3 XP_016881096.1
XM_017025608.1 4113 UTR 3 XP_016881097.1
XM_017025609.1 4113 UTR 3 XP_016881098.1

View Full Product Details