Product Details

SNP ID
rs77492032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:25067053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAAAGGAAGTAGGAAGAGAAG[A/G]AAACTTCTGTAATCCCACAGAGAGC
Phenotype
MIM: 610974
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF521 PubMed Links

Gene Details

Gene
ZNF521
Gene Name
zinc finger protein 521
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308225.1 Intron NP_001295154.1
NM_015461.2 Intron NP_056276.1
XM_011525909.2 Intron XP_011524211.1
XM_011525910.2 Intron XP_011524212.1
XM_011525911.1 Intron XP_011524213.1
XM_017025697.1 Intron XP_016881186.1
XM_017025698.1 Intron XP_016881187.1

View Full Product Details