Product Details

SNP ID
rs78247107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37885077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCACACAGTTCTGCCATGGGAGG[A/G]GTACCGCACCCTGGAACTCCTGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR87 PubMed Links

Gene Details

Gene
WDR87
Gene Name
WD repeat domain 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291088.1 8848 Missense Mutation CCC,CTC P2865L NP_001278017.1
NM_031951.4 8848 Missense Mutation CCC,CTC P2826L NP_114157.4
XM_011527360.2 8848 Missense Mutation CCC,CTC P2865L XP_011525662.1

View Full Product Details