Product Details

SNP ID
rs80248244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38351074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTCAAGCCCACATTCAGTGGG[A/C]AAGATCTTTCCCAAGAATCACCACA
Phenotype
MIM: 613452
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CATSPERG PubMed Links

Gene Details

Gene
CATSPERG
Gene Name
cation channel sperm associated auxiliary subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021185.4 Intron NP_067008.3
XM_005259114.4 Intron XP_005259171.1
XM_005259115.4 Intron XP_005259172.1
XM_005259116.2 Intron XP_005259173.1
XM_006723308.3 Intron XP_006723371.1
XM_006723310.2 Intron XP_006723373.1
XM_006723311.2 Intron XP_006723374.1
XM_011527174.2 Intron XP_011525476.1
XM_011527175.2 Intron XP_011525477.1
XM_011527176.2 Intron XP_011525478.1
XM_011527177.2 Intron XP_011525479.1
XM_011527178.2 Intron XP_011525480.1
XM_011527179.1 Intron XP_011525481.1
XM_011527182.2 Intron XP_011525484.1
XM_017027053.1 Intron XP_016882542.1
XM_017027054.1 Intron XP_016882543.1
XM_017027055.1 Intron XP_016882544.1

View Full Product Details