Product Details

SNP ID
rs77624079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39515450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTCCCCACTCCCGCTCTGGCC[C/T]GGATCCCCCGTCTGGTTCCGCCTCC
Phenotype
MIM: 602768 MIM: 607919
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLL3 PubMed Links

Gene Details

Gene
DLL3
Gene Name
delta like canonical Notch ligand 3
There are no transcripts associated with this gene.

Gene
SELV
Gene Name
selenoprotein V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182704.1 338 Missense Mutation CGG,TGG R80W NP_874363.1

View Full Product Details