Product Details

SNP ID
rs77151878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40030029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTATGGTCTACAATATTTTCA[A/G]TTTAACAATGGGTTTATCTAGATAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF546 PubMed Links

Gene Details

Gene
ZNF546
Gene Name
zinc finger protein 546
There are no transcripts associated with this gene.

Gene
ZNF780B
Gene Name
zinc finger protein 780B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005851.2 6895 UTR 3 NP_001005851.1
XM_005258590.4 6895 Intron XP_005258647.1
XM_005258591.4 6895 Intron XP_005258648.1
XM_005258592.3 6895 Intron XP_005258649.1
XM_005258593.2 6895 Intron XP_005258650.1
XM_005258595.3 6895 Intron XP_005258652.1
XM_006723072.3 6895 Intron XP_006723135.1
XM_011526593.2 6895 Intron XP_011524895.1
XM_017026425.1 6895 Intron XP_016881914.1
XM_017026426.1 6895 Intron XP_016881915.1
XM_017026427.1 6895 Intron XP_016881916.1

View Full Product Details