Product Details

SNP ID
rs78772583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40070590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTAAAATTGAGAAAATTCTTTTT[A/G]TAAGTGAATTCAGTATGTTCACTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF780A PubMed Links

Gene Details

Gene
ZNF780A
Gene Name
zinc finger protein 780A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010880.2 6080 Intron NP_001010880.2
NM_001142577.1 6080 Intron NP_001136049.1
NM_001142578.1 6080 Intron NP_001136050.1
NM_001142579.1 6080 UTR 3 NP_001136051.1
XM_005258773.2 6080 UTR 3 XP_005258830.1
XM_006723150.3 6080 UTR 3 XP_006723213.1
XM_011526771.2 6080 UTR 3 XP_011525073.1
XM_011526772.2 6080 UTR 3 XP_011525074.1
XM_017026611.1 6080 UTR 3 XP_016882100.1
XM_017026612.1 6080 UTR 3 XP_016882101.1
XM_017026613.1 6080 UTR 3 XP_016882102.1
XM_017026614.1 6080 UTR 3 XP_016882103.1
XM_017026615.1 6080 UTR 3 XP_016882104.1
XM_017026616.1 6080 UTR 3 XP_016882105.1
XM_017026617.1 6080 UTR 3 XP_016882106.1
XM_017026618.1 6080 UTR 3 XP_016882107.1
XM_017026619.1 6080 UTR 3 XP_016882108.1

View Full Product Details